missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferric Ammonium Sulfate TS, 8% (w/v), Ricca Chemical
Supplier: Ricca Chemical Company 305832
Specifications
Ferric Ammonium Sulfate | |
Test Solution | |
91.0,7.26 | |
FeH28NO20S2 | |
LCPUDZUWZDSKMX-UHFFFAOYSA-K | |
azanium;iron(3+);disulfate;dodecahydrate | |
197097 | |
Aqueous Solution | |
Liquid |
Amber | |
7783-83-7,7732-18-5 | |
94.0,7.56 | |
ammonium ferric sulfate, ferric ammonium sulfate dodecahydrate, iron ammonium sulfate dodecahydrate, sulfuric acid, ammonium iron 3+ salt 2:1:1 , dodecahydrate, ferric ammonium sulfate, acs, azanium iron 3+ disulfate dodecahydrate, ferric iron ammonium dodecahydrate disulfate, azanium iron +3 cation disulfate dodecahydrate, ammonium iron iii sulfate dodecahydrate, acs 100g | |
[NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3] | |
482.176 | |
Natural Poly Bottle | |
1 L | |
8% (w/v) |
Chemical Identifiers
7783-83-7 | |
482.176 | |
ammonium ferric sulfate, ferric ammonium sulfate dodecahydrate, iron ammonium sulfate dodecahydrate, sulfuric acid, ammonium iron 3+ salt 2:1:1 , dodecahydrate, ferric ammonium sulfate, acs, azanium iron 3+ disulfate dodecahydrate, ferric iron ammonium dodecahydrate disulfate, azanium iron +3 cation disulfate dodecahydrate, ammonium iron iii sulfate dodecahydrate, acs 100gShow More | |
azanium;iron(3+);disulfate;dodecahydrate |
FeH28NO20S2 | |
LCPUDZUWZDSKMX-UHFFFAOYSA-K | |
197097 | |
[NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3] |