missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferric Ammonium Sulfate TS, 8% (w/v), Ricca Chemical
$120.80 - $338.17
Chemical Identifiers
| CAS | 7783-83-7 |
|---|---|
| Molecular Formula | FeH28NO20S2 |
| Molecular Weight (g/mol) | 482.176 |
| InChI Key | LCPUDZUWZDSKMX-UHFFFAOYSA-K |
| Synonym | ammonium ferric sulfate, ferric ammonium sulfate dodecahydrate, iron ammonium sulfate dodecahydrate, sulfuric acid, ammonium iron 3+ salt 2:1:1 , dodecahydrate, ferric ammonium sulfate, acs, azanium iron 3+ disulfate dodecahydrate, ferric iron ammonium dodecahydrate disulfate, azanium iron +3 cation disulfate dodecahydrate, ammonium iron iii sulfate dodecahydrate, acs 100g |
| PubChem CID | 197097 |
| IUPAC Name | azanium;iron(3+);disulfate;dodecahydrate |
| SMILES | [NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
3058-4
|
Ricca Chemical Company
30584 |
120 mL | Natural Poly Bottle |
Each for $120.80
|
|
||||
|
3058-16
|
Ricca Chemical Company
305816 |
500 mL | Natural Poly Bottle |
Each for $207.40
|
|
||||
|
305832
|
Ricca Chemical Company
305832 |
1 L | Natural Poly Bottle |
Each for $338.17
|
|
||||
Chemical Identifiers
| 7783-83-7 | |
| 482.176 | |
| ammonium ferric sulfate, ferric ammonium sulfate dodecahydrate, iron ammonium sulfate dodecahydrate, sulfuric acid, ammonium iron 3+ salt 2:1:1 , dodecahydrate, ferric ammonium sulfate, acs, azanium iron 3+ disulfate dodecahydrate, ferric iron ammonium dodecahydrate disulfate, azanium iron +3 cation disulfate dodecahydrate, ammonium iron iii sulfate dodecahydrate, acs 100g | |
| azanium;iron(3+);disulfate;dodecahydrate |
| FeH28NO20S2 | |
| LCPUDZUWZDSKMX-UHFFFAOYSA-K | |
| 197097 | |
| [NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3] |
Specifications
| Amber | |
| 91.0,7.26 | |
| FeH28NO20S2 | |
| LCPUDZUWZDSKMX-UHFFFAOYSA-K | |
| azanium;iron(3+);disulfate;dodecahydrate | |
| 197097 | |
| Laboratory | |
| Aqueous Solution | |
| 120 mL | |
| 8% (w/v) |
| 7783-83-7 , 7732-18-5 | |
| 94.0,7.56 | |
| ammonium ferric sulfate, ferric ammonium sulfate dodecahydrate, iron ammonium sulfate dodecahydrate, sulfuric acid, ammonium iron 3+ salt 2:1:1 , dodecahydrate, ferric ammonium sulfate, acs, azanium iron 3+ disulfate dodecahydrate, ferric iron ammonium dodecahydrate disulfate, azanium iron +3 cation disulfate dodecahydrate, ammonium iron iii sulfate dodecahydrate, acs 100g | |
| [NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3] | |
| 482.176 | |
| 8% | |
| Natural Poly Bottle | |
| ∼2.5 | |
| Liquid | |
| Ferric Ammonium Sulfate, Test Solution |