missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fehling A solution
CAS: 7758-98-7 | CuO4S | 159.602 g/mol
Supplier: Thermo Scientific Chemicals 270800025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Greener Choice Claims
- Environmental benefits include:
- Renewable Energy
Specifications
Fehling A solution | |
7758-98-7 | |
CuSO4 | |
1.04g/mL | |
Solubility in water: soluble. | |
[O-]S(=O)(=O)[O-].[Cu+2] | |
159.602 | |
CHEBI:23414 | |
Plastic bottle | |
Solution |
Blue | |
CuO4S | |
MFCD00010981 | |
copper sulfate, copper ii sulfate, cupric sulfate anhydrous, copper sulphate, copper 2+ sulfate, blue stone, copper monosulfate, copper ii sulfate, anhydrous, hylinec, trinagle | |
ARUVKPQLZAKDPS-UHFFFAOYSA-L | |
copper;sulfate | |
24462 | |
159.61 | |
2.5 L |
Chemical Identifiers
7758-98-7 | |
159.602 | |
ARUVKPQLZAKDPS-UHFFFAOYSA-L | |
24462 | |
copper;sulfate |
CuO4S | |
MFCD00010981 | |
copper sulfate, copper ii sulfate, cupric sulfate anhydrous, copper sulphate, copper 2+ sulfate, blue stone, copper monosulfate, copper ii sulfate, anhydrous, hylinec, trinagleShow More | |
CHEBI:23414 | |
[O-]S(=O)(=O)[O-].[Cu+2] |
RUO – Research Use Only