missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fehling A solution
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Blue |
| Linear Formula | CuSO4 |
| Density | 1.04g/mL |
| Solubility Information | Solubility in water: soluble. |
| Formula Weight | 159.61 |
| Packaging | Plastic bottle |
| Quantity | 1 L |
| Physical Form | Solution |
| Chemical Name or Material | Fehling A solution |
RUO – Research Use Only