missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ethyle5-(5-bromo-2-thienyl)-3-isoxazolecarboxylate, 97%, Thermo Scientific™

Chemical Identifiers
CAS | 423768-50-7 |
---|---|
Molecular Formula | C10H8BrNO3S |
Molecular Weight (g/mol) | 302.142 |
MDL Number | MFCD03407336 |
InChI Key | OIZZMDOBYXRSNQ-UHFFFAOYSA-N |
Synonym | ethyl 5-5-bromo-2-thienyl-3-isoxazolecarboxylate, ethyl 5-5-bromothiophen-2-yl-1,2-oxazole-3-carboxylate, ethyl 5-5-bromothiophen-2-yl isoxazole-3-carboxylateShow More |
PubChem CID | 2776548 |
IUPAC Name | ethyl 5-(5-bromothiophen-2-yl)-1,2-oxazole-3-carboxylate |
SMILES | CCOC(=O)C1=NOC(=C1)C2=CC=C(S2)Br |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
CC23210CB
|
Thermo Scientific Chemicals
CC23210CB |
250mg |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
CC23210DA
|
Thermo Scientific Chemicals
CC23210DA |
1g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
CC23210DE
|
Thermo Scientific Chemicals
CC23210DE |
5g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
423768-50-7 | |
302.142 | |
OIZZMDOBYXRSNQ-UHFFFAOYSA-N | |
2776548 | |
CCOC(=O)C1=NOC(=C1)C2=CC=C(S2)Br |
C10H8BrNO3S | |
MFCD03407336 | |
ethyl 5-5-bromo-2-thienyl-3-isoxazolecarboxylate, ethyl 5-5-bromothiophen-2-yl-1,2-oxazole-3-carboxylate, ethyl 5-5-bromothiophen-2-yl isoxazole-3-carboxylateShow More | |
ethyl 5-(5-bromothiophen-2-yl)-1,2-oxazole-3-carboxylate |
Specifications
423768-50-7 | |
100.0 | |
Amber glass bottle | |
MFCD03407336 | |
ethyl 5-5-bromo-2-thienyl-3-isoxazolecarboxylate, ethyl 5-5-bromothiophen-2-yl-1,2-oxazole-3-carboxylate, ethyl 5-5-bromothiophen-2-yl isoxazole-3-carboxylate | |
CCOC(=O)C1=NOC(=C1)C2=CC=C(S2)Br | |
302.142 | |
302.15 | |
ethyle5-(5-bromo-2-thienyl)-3-isoxazolecarboxylate |
96.5 | |
97% | |
C10H8BrNO3S | |
250mg | |
OIZZMDOBYXRSNQ-UHFFFAOYSA-N | |
ethyl 5-(5-bromothiophen-2-yl)-1,2-oxazole-3-carboxylate | |
2776548 | |
97% |