missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ethyle5-(5-bromo-2-thienyl)-3-isoxazolecarboxylate, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC23210DA
Specifications
| ethyle5-(5-bromo-2-thienyl)-3-isoxazolecarboxylate | |
| 96.5 | |
| 97% | |
| C10H8BrNO3S | |
| 1g | |
| OIZZMDOBYXRSNQ-UHFFFAOYSA-N | |
| ethyl 5-(5-bromothiophen-2-yl)-1,2-oxazole-3-carboxylate | |
| 2776548 | |
| 97% |
| 423768-50-7 | |
| 100.0 | |
| Amber glass bottle | |
| MFCD03407336 | |
| ethyl 5-5-bromo-2-thienyl-3-isoxazolecarboxylate, ethyl 5-5-bromothiophen-2-yl-1,2-oxazole-3-carboxylate, ethyl 5-5-bromothiophen-2-yl isoxazole-3-carboxylate | |
| CCOC(=O)C1=NOC(=C1)C2=CC=C(S2)Br | |
| 302.142 | |
| 302.15 |
Chemical Identifiers
| 423768-50-7 | |
| 302.142 | |
| OIZZMDOBYXRSNQ-UHFFFAOYSA-N | |
| 2776548 | |
| CCOC(=O)C1=NOC(=C1)C2=CC=C(S2)Br |
| C10H8BrNO3S | |
| MFCD03407336 | |
| ethyl 5-5-bromo-2-thienyl-3-isoxazolecarboxylate, ethyl 5-5-bromothiophen-2-yl-1,2-oxazole-3-carboxylate, ethyl 5-5-bromothiophen-2-yl isoxazole-3-carboxylate | |
| ethyl 5-(5-bromothiophen-2-yl)-1,2-oxazole-3-carboxylate |