missing translation for 'onlineSavingsMsg'
Learn More

ethyle5-(5-bromo-2-thienyl)-3-isoxazolecarboxylate, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC23210CB

 View more versions of this product

Catalog No. CC23210CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

ethyle5-(5-bromo-2-thienyl)-3-isoxazolecarboxylate
96.5
97%
C10H8BrNO3S
250mg
OIZZMDOBYXRSNQ-UHFFFAOYSA-N
ethyl 5-(5-bromothiophen-2-yl)-1,2-oxazole-3-carboxylate
2776548
97%
423768-50-7
100.0
Amber glass bottle
MFCD03407336
ethyl 5-5-bromo-2-thienyl-3-isoxazolecarboxylate, ethyl 5-5-bromothiophen-2-yl-1,2-oxazole-3-carboxylate, ethyl 5-5-bromothiophen-2-yl isoxazole-3-carboxylate
CCOC(=O)C1=NOC(=C1)C2=CC=C(S2)Br
302.142
302.15

Chemical Identifiers

423768-50-7
302.142
OIZZMDOBYXRSNQ-UHFFFAOYSA-N
2776548
CCOC(=O)C1=NOC(=C1)C2=CC=C(S2)Br
C10H8BrNO3S
MFCD03407336
ethyl 5-5-bromo-2-thienyl-3-isoxazolecarboxylate, ethyl 5-5-bromothiophen-2-yl-1,2-oxazole-3-carboxylate, ethyl 5-5-bromothiophen-2-yl isoxazole-3-carboxylate
ethyl 5-(5-bromothiophen-2-yl)-1,2-oxazole-3-carboxylate
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Promotions

Promotions