Learn More
Eppendorf Referenceā„¢ 2, Mechanical Single-Channel Pipette
GREENER_CHOICE
Minimize risk of aerosol contamination and reduce thumb strain with these variable volume pipettes. Eppendorfā„¢ Referenceā„¢ 2 Variable Volume Pipettes are ideal for pipetting and mixing liquids, filling of plates, gels and reaction vessels, and phase extraction and removal of supernatants.
Supplier:  Eppendorf 4924000088
 View more versions of this product
Catalog No. 05-412-428
Only null left
Explore available promotions
Description
Description
  • Integrated pipetting system with internal RFID technology
  • Serial number is printed on multiple components of pipette
  • Color coded and volume labeled pipette for quick identification of volume size/tip size
  • Round upper part of pipette makes it possible to work in every position
  • Four-digit display for exact, quick and secure volume setting (volume lock included)
  • Spring-loaded tip cone reduces effort during operation/ejection
  • Sturdy upper handle, movable parts and external edges made from stainless-steel make pipette robust
  • High flexibility with ability to remove single cones
  • Ensures continuous identical pipet alignment during work process
  • Fully autoclavable pipet with smooth surface and few rough edges
  • Single-button operation
  • Has Spring-loaded tip cone, movable parts, stainless steel external edges with customizable labeling area
  • Ergonomic handling with reduced operating effort
  • Simple and quick tip ejection with active aerosol reduction
  • Reduce repetitive strain injuries by not having to move your thumb sideways
  • Haptic feedback before tip ejection
  • High precision and accuracy provides reliable pipetting results
  • User friendly secondary adjustment
Greener Choice Claims
  • Environmental benefits include:
    • Ozone-Safe or Safe for the Ozone Layer
    • Non-Toxic
    • Renewable Energy
    • Free-of a substance that causes environmental harm
Specifications
Specifications
Autoclavable
Pipette Tip 1000 μL
Reference 2
100 to 1000 μL
epT.I.P.S.ā„¢ Box 2.0 with 96 Tips
Blue
Blue
1
Pipette
1000 μL pipette tips
1 μL
Product Suggestions
Product Suggestions