missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf Reference™ 2, Mechanical Single-Channel Pipette
GREENER_CHOICE

Catalog No. p-4760726
Change view
Click to view available options
Volume (Metric):
0.1 to 2.5 μL
0.25 to 2.5 mL
0.5 to 10 μL
0.5 to 5 mL
1 to 10 mL
10 to 100 μL
100 to 1000 μL
2 to 20 μL
20 to 200 μL
30 to 300 μL
Color:
Blue
Dark Gray
Light Gray
Medium Gray
Orange
Red
Turquoise
Violet
Yellow
Increments:
0.002 μL
0.01 μL
0.02 μL
0.1 μL
0.2 μL
1 μL
2 μL
5 μL
10 μL
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Color Increments
05412422 0.5 to 10 μL Medium Gray 0.01 μL
05412421 0.1 to 2.5 μL Dark Gray 0.002 μL
05412423 2 to 20 μL Light Gray 0.02 μL
05412424 2 to 20 μL Yellow 0.02 μL
05412425 10 to 100 μL Yellow 0.1 μL
05412426 20 to 200 μL Yellow 0.2 μL
05412427 30 to 300 μL Orange 0.2 μL
05412428 100 to 1000 μL Blue 1 μL
05412430 0.5 to 5 mL Violet 5 μL
05412429 0.25 to 2.5 mL Red 2 μL
05412431 1 to 10 mL Turquoise 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 05412422 Supplier Eppendorf Supplier No. 4924000029
Only null left

Minimize risk of aerosol contamination and reduce thumb strain with these variable volume pipettes. Eppendorf™ Reference™ 2 Variable Volume Pipettes are ideal for pipetting and mixing liquids, filling of plates, gels and reaction vessels, and phase extraction and removal of supernatants.

  • Integrated pipetting system with internal RFID technology
  • Serial number is printed on multiple components of pipette
  • Color coded and volume labeled pipette for quick identification of volume size/tip size
  • Round upper part of pipette makes it possible to work in every position
  • Four-digit display for exact, quick and secure volume setting (volume lock included)
  • Spring-loaded tip cone reduces effort during operation/ejection
  • Sturdy upper handle, movable parts and external edges made from stainless-steel make pipette robust
  • High flexibility with ability to remove single cones
  • Ensures continuous identical pipet alignment during work process
  • Fully autoclavable pipet with smooth surface and few rough edges
  • Single-button operation
  • Has Spring-loaded tip cone, movable parts, stainless steel external edges with customizable labeling area
  • Ergonomic handling with reduced operating effort
  • Simple and quick tip ejection with active aerosol reduction
  • Reduce repetitive strain injuries by not having to move your thumb sideways
  • Haptic feedback before tip ejection
  • High precision and accuracy provides reliable pipetting results
  • User friendly secondary adjustment

Specifications

Autoclavable Autoclavable
Color Medium Gray
For Use With Pipette Tip 20 μL
Number of Channels 1
Product Line Reference 2
Type Pipette
Volume (Metric) 0.5 to 10 μL
For Use With (Equipment) 20 μL pipette tips
Includes epT.I.P.S.™ Box 2.0 with 96 Tips
Increments 0.01 μL