missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Upstate Endothelial Cell Growth Supplements
GREENER_CHOICE

Designed for use as a cell culture media supplement

$447.00 - $447.00

Specifications

Content And Storage 2 years at 4°C; rehydrated: 3 months at -20°C
For Use With (Application) Cell Culture
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
02101MI
View Documents
MilliporeSigma™
02101
50 mg
Each for $447.00
Only null left
 
Description

Description

ECGS is mitogenic for many types of cells under reduced or serum-free conditions, such as mammalian, avian and human endothelial cells, smooth muscle cells, keratinocytes, melanocytes and hybridomas; ECGS often fully substitutes for feeder layers in culture of fastidious cells.
Specifications

Specifications

2 years at 4°C; rehydrated: 3 months at -20°C
Cell Culture
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'