MilliporeSigma™ SeQuant™ ZIC™-cHILIC Metal-free HPLC PEEK Columns
PEEK coated HPLC column
Supplier: MilliporeSigma™ 1506610001
Description
SeQuant™ ZIC™-cHILIC is designed for improved separation of polar hydrophilic compounds that have proved difficult to separate by reversed-phase HPLC or previous types of HILIC columns. The orientation of its zwitterionic functional group provides more accessible positive charges, which results in a complementary selectivity with more retention particularly for organic acids and other negatively charged compounds. Especially suitable for LC-MS thanks to an extraordinary high column stability and a minimized column bleed.Specifications
4.6 mm | |
100 Å | |
1/Pk. |
100 mm | |
100 AÌ | |
Column |