Learn More
DuPont™ Tychem™ QC Sleeved Apron
GREENER_CHOICE
Knee-length apron/gown design with attached sleeves
$271.96 - $293.72
Specifications
Seam Type Bound
View More Specs
Products 2
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
19-067-019
View Documents
DuPont™
QC275BYL3X002500
3X-Large
Case of 25 Each for $271.96
Only null left
19-067-020
View Documents
DuPont™
QC275BYL4X002500
4X-Large
Case of 25 Each for $293.72
Only null left
Description
Description
Tychem™ QC fabric provides at least 30 minutes of protection against 42 chemical challenges. It is used for light splash protection in a variety of industrial environments, including petroleum refining, pulp and paper manufacturing, food processing, chemical processing, and pharmaceutical manufacturing.
  • Lightweight and durable fabric
  • Provides enhanced frontal protection
  • Tightly sewn seam is covered with garment fabric to reinforce seam and reduce potential for particle penetration
  • Open back for wearer comfort
  • Bound seams around neck opening with snap closure
  • Elastic opening for tighter fit at wrist
  • Ties at waist and elastic wrists
  • Yellow colored apron is 44 in. long for neck-to-knee coverage
Specifications
Specifications
Bound
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'