Learn More
DuPont™ Tychem™ QC Sleeved Apron
Knee-length apron/gown design with attached sleeves
Supplier:  DuPont™ QC275BYL3X002500
 View more versions of this product
Catalog No. 19-067-019
Only null left
Explore available promotions
Description
Description
Tychem™ QC fabric provides at least 30 minutes of protection against 42 chemical challenges. It is used for light splash protection in a variety of industrial environments, including petroleum refining, pulp and paper manufacturing, food processing, chemical processing, and pharmaceutical manufacturing.
  • Lightweight and durable fabric
  • Provides enhanced frontal protection
  • Tightly sewn seam is covered with garment fabric to reinforce seam and reduce potential for particle penetration
  • Open back for wearer comfort
  • Bound seams around neck opening with snap closure
  • Elastic opening for tighter fit at wrist
  • Ties at waist and elastic wrists
  • Yellow colored apron is 44 in. long for neck-to-knee coverage
Greener Choice Claims
  • Environmental benefits include:
    • Recyclable
Specifications
Specifications
3X-Large
Bound
Product Suggestions
Product Suggestions