Learn More
DuPont™ Tychem™ 2000 Sleeved Apron
QC Sleeved Aprons offer added chemical protection from numerous light splash situations for a variety of industries.
Supplier: DuPont™ QC275BYL4X002500
Avertissement:
Should not be worn around open flame, heat source, sparks or ignition sources. If the garment is damaged, do not use.
Description
- Knee-length apron/gown design with attached sleeves
- Available in multiple sizes
- Provides enhanced frontal protection
- Tightly sewn seam is covered with garment fabric to reinforce seam and reduce potential for particle penetration
- Open back for wearer comfort
- Bound seams around neck opening with snap closure
- Ties at waist
- Elastic opening for tighter fit at wrist
- 44 in. long for neck-to-knee coverage
- Manufactured under specifications that do not contain natural rubber latex
Spécifications
| 4X-Large | |
| Yellow | |
| Elastic | |
| Bound |
| Tychem | |
| Waist Ties | |
| 10 mil |