Learn More
Corning™ BioCoat™ Matrigel™ Invasion Chamber with Corning™ Matrigel Matrix
Provide rapid and reproducible quantification of cell invasion in vitro. Corning™ BioCoat™ Matrigel™ Invasion Chamber with Corning Matrigel Matrix™ cell culture inserts are pre-coated with extracellular matrix proteins for applications requiring a protein-coated cell surface, such as cell adhesion, growth, invasion, migration and differentiation.
Supplier: Corning™ 354481
Description
Corning BioCoat cell culture inserts are pre-coated with extracellular matrix proteins for applications requiring a protein-coated cell surface, such as cell adhesion, growth, invasion, migration and/or differentiation.
Coatings include Corning Matrigel™ matrix, Fibronectin, Collagen, and Laminin.
- Environmental benefits include:
- Source Reduction
Specifications
6 | |
PET | |
Studies in which minimal interference by growth factors is desirable | |
6/Pk. | |
Matrigel™ Matrix |
4.2 cm2 | |
8 μm | |
4 | |
24/Cs. |