Learn More
Contec™ QuickConnect™ Mop Head Frames
Featuring a quick-connect button for easy attachment to all sizes and styles of Contec™ Quick Connect™ handles. QuickConnect™ Mop Head Frames are specifically designed for use with the Contec™ family of mopping systems.
Supplier:  Contec™ 2685
 View more versions of this product
Catalog No. 19-033-780
Only null left
Explore available promotions
Description
Description
Featuring a quick-connect button for easy attachment to all sizes and styles of Contec™ Quick Connect™ handles. Contec™ QuickConnect™ Mop Head Frames are specifically designed for use with the Contec™ family of mopping systems.
  • Offer maximum flexibility for cleaning in critical environments
  • Interchangeable frames for Contec Mopping Systems
  • Autoclavable
  • Capped end and quick-connect feature for easy attachment
Specifications
Specifications
Mop Head Frame
Non-sterile
Autoclavable
QuickConnect Mop Handles
Designed for Use in Pharmaceutical and Medical Device Manufacturing Facilities, or Other Similar Critical or Controlled Environments.
7.5 in.
19 cm
Reusable
EasyCurve QuickConnect Mop Head Frame
Stainless Steel
Product Suggestions
Product Suggestions