missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution (Phosphate), pH 7, BAKER ANALYZED™ Reagent, J.T. Baker™

High quality chemicals for laboratory and specialized industrial use

$4685.11 - $5276.00

Specifications

Chemical Name or Material Buffer Solution (Phosphate), pH 7
Specific Gravity 1L = 1.00kg
Grade BAKER ANALYZED™ Reagent
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
02002488
View Documents
Avantor J.T.Baker
560801
500 mL Poly Bottle
Case of 12 Each for $4,685.11
Only null left
 
02002487
View Documents
Avantor J.T.Baker
560803
4 L Cubetainer
Case of 4 Each for $5,276.00
Only null left
 
Description

Description

Specifications

Specifications

Buffer Solution (Phosphate), pH 7
1L = 1.00kg
BAKER ANALYZED™ Reagent
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'