missing translation for 'onlineSavingsMsg'
Learn More

Brady™ SPC Re-Form Universal Sorbent Pads
GREENER_CHOICE

Unique composition absorbs up to 50% more than traditional meltblown polypropylene sorbents

$106.99 - $146.37

Specifications

Certifications/Compliance NFPA Class A
Color Gray
For Use With (Application) Ideal for handling foot traffic and wiping applications, indoor use or dry-weather outdoor use
Dimensions (L x W) 19 x 15 in. (48.2 x 38 cm)
Length (English) 19 in.
View More Specs

Products 3
Catalog Number Mfr. No. Capacity (English) Weight Capacity Type Price Quantity  
Catalog Number Mfr. No. Capacity (English) Weight Capacity Type Price Quantity  
19-190-053
View Documents
Brady™
RFP100
40 gal. Heavyweight Re-Form Plus, Medium-Linting N/A
Only null left
 
19-190-054
View Documents
Brady™
RFP300
32 gal. Medium-weight Re-Form Plus, Medium-Linting N/A
Only null left
 
19-190-055
View Documents
Brady™
RFP500
24 gal. Lightweight Re-Form Plus, Medium-Linting N/A
Only null left
 
Description

Description

  • For basic drips and spills
  • Netted construction for extra durability
  • Indoor use or dry-weather outdoor use
  • General-purpose sorbent (not for use with highly agressive acids and caustics)
  • Re-Form Plus pads feature perforated spunbound coverstock on one side that withstands foot traffic
  • All pads measure 15 x 19 in. (38.1 x 48.3cm)
  • Color: Gray

Specifications

Specifications

NFPA Class A
Ideal for handling foot traffic and wiping applications, indoor use or dry-weather outdoor use
19 in.
15 in.
Gray
19 x 15 in. (48.2 x 38 cm)
48.2 cm
38 cm
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'