Learn More
Brady™ Safety First Emergency Shower/Eye Bath Sign
Safety First Emergency Shower/Eye Bath Sign from Brady help to manage and sustain a clean and safe workplace.
Supplier:  Brady™ 85046
 View more versions of this product
Catalog No. 19-036023
Only null left
Explore available promotions
Description
Description
  • Self-Sticking Polyester (B-302) is an Indoor/Outdoor high performance pressure sensitive sign that is surface printed polyester with a polyester overlaminate.
  • Plastic signs (B-401) are ideal for use in light duty industrial, utility, commercial and institutional environments.
Specifications
Specifications
OSHA 1910.145, PSTC-33
1 Ea.
Surface Mount
10 in.
-40°F
14 in.
-40°C to 110°C
10 x 14 in.
First Aid Sign
Polyester
Permanent Acrylic
Used for safety signs, identification of fire alarms, extinguishers and emergency exits
230°F
0.006 to 0.01 in.
35.56 cm
-40°F to 230°F
Emergency Shower Eye Bath
White
Product Suggestions
Product Suggestions