Learn More
Brady™ Large Horizontal No Smoking Signs
Large Horizontal No Smoking Signs from Brady help to manage and sustain a clean and safe workplace.
Supplier:  Brady™ 88425
 View more versions of this product
Catalog No. 19-036-204
Only null left
Explore available promotions
Description
Description
  • Self-Sticking Polyester (B-302) is an Indoor/Outdoor high performance pressure sensitive sign that is surface printed polyester with a polyester overlaminate.
  • Plastic signs (B-401) are ideal for use in light duty industrial, utility, commercial and institutional environments.
  • Extremely durable (B-120) Premium Fiberglass will not chip, fade, rust, shatter or peel for up to 25 years.
Specifications
Specifications
OSHA
1 Ea.
Safety sign, identification of fire alarm, extinguisher and emergency exit
230°F
0.006 to 0.01 in.
25.4 cm
-40°F to 230°F
No Smoking
Red on White
Polyester
Permanent Acrylic
7 in.
-40°F
10 in.
-40°C to 110°C
7 x 10 in.
No Smoking Sign
Product Suggestions
Product Suggestions