Learn More
Brady™ Exit and Directional Signs
Exit and Directional Signs from Brady guide traffic and lead employees to safety during emergency situations.
Supplier:  Brady™ 84685
 View more versions of this product
Catalog No. 19-015751
Only null left
Explore available promotions
Description
Description
  • Self-Sticking Polyester (B-302) is an Indoor/Outdoor high performance pressure sensitive sign that is surface printed polyester with a polyester overlaminate.
  • Plastic signs (B-401) are ideal for use in light duty industrial, utility, commercial and institutional environments.
  • Extremely durable (B-120) Premium Fiberglass will not chip, fade, rust, shatter or peel for up to 25 years.
  • Durable Non-Reflective Aluminum (B-555) is a rigid aluminum panel with protected graphics designed for use in industrial, utility, commercial and institutional environments.
Greener Choice Claims
  • Environmental benefits include:
    • Recyclable
Specifications
Specifications
ANSI Z535, OSHA 1910.145
Polyester
17.7 H x 25.4 W x 0.02 cm D
Used for safety signs, identification of fire alarms, extinguishers and emergency exits
17.7 cm
-40°F
10 in.
-40°C to 110°C
7 x 10 in.
No Exit Sign
Industrial
1 Ea.
Permanent Acrylic
7 in.
230°F
0.006 to 0.01 in.
25.4 cm
-40°F to 230°F
No Exit
Red on White
Product Suggestions
Product Suggestions