Learn More
Thermo Scientificâ„¢ Bottom-of-the-Bottleâ„¢ Solvent Filters
Protect your HPLC system with Thermo Scientificâ„¢ Bottom-of-the-Bottleâ„¢ Solvent Filters.
$93.53 - $143.31
Specifications
Type Filter Kit
View More Specs
Products 3
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity  
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity  
03-052-397
View Documents
Thermo Scientificâ„¢
A302
Fit 1/16 in. OD Tube to 1/8 in. OD Plastic Tubing
Each for $93.53
Only null left
03052398
View Documents
Thermo Scientificâ„¢
A302A
Fit to 1/8 in. OD Plastic Tubing Using 1/8 in. Polypropylene Nut
Each for $128.91
Only null left
03-170-305
View Documents
Thermo Scientificâ„¢
A437
1/8 in. OD Tubing
Each for $143.31
Only null left
Description
Description
Protect your HPLC system from contamination with Thermo Scientific™ Bottom-of-the-Bottle™ Solvent Filters. The flat design ensures efficient draw. They are made from 100% PEEK polymer and include two 10μm filters. A helium sparge port and frit are incorporated for optional use.
  • Efficient draw
  • 100% PTFE polymer, including 2μm filter
  • Built-in helium sparge port and frit
  • Stainless-steel 10μm inlet filters for longer lifetime
Specifications
Specifications
Filter Kit
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.