missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Amicon™ Ultra-15 Centrifugal Filter Units
Achieve both high sample recovery and fast sample processing. EMD Amicon™ Ultra Centrifugal Filter Units feature regenerated cellulose membranes that are ideal for protein & nucleic acid purification, concentration and desalting.
$165.00 - $1690.00
Specifications
Disposable | Single-use |
---|---|
Material (Membrane) | High-recovery Ultracel-PL Regenerated Cellulose |
Sterility | Non-sterile |
Filtration Area | 7.6 cm2 |
For Use With (Application) | Protein purification, protein concentration and desalting |
Catalog Number | Mfr. No. | MWCO | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | MWCO | Quantity | Price | Quantity | |||||
UFC900308
|
MilliporeSigma™
UFC900308 |
3 kDa | 8Pack |
Each for $166.00
|
|
|||||
UFC900396
|
MilliporeSigma™
UFC900396 |
3 kDa | 96 Pack |
Each for $1,690.00
|
|
|||||
UFC901008
|
MilliporeSigma™
UFC901008 |
10 kDa | 8Pack |
Each for $165.00
|
|
|||||
UFC901024
![]() |
MilliporeSigma™
UFC901024 |
10 kDa | 24Pack |
Each for $435.00
|
![]() |
|||||
UFC901096
|
MilliporeSigma™
UFC901096 |
10 kDa | Pack of 96 |
Each for $1,690.00
|
|
|||||
UFC903008
|
MilliporeSigma™
UFC903008 |
30 kDa | 8Pack |
Each for $166.00
|
|
|||||
UFC903024
|
MilliporeSigma™
UFC903024 |
30 kDa | 24Pack |
Each for $435.00
|
|
|||||
UFC903096
|
MilliporeSigma™
UFC903096 |
30 kDa | 96Pack |
Each for $1,690.00
|
|
|||||
UFC905008
|
MilliporeSigma™
UFC905008 |
50 kDa | 8Pack |
Each for $165.00
|
|
|||||
UFC905096
|
MilliporeSigma™
UFC905096 |
50 kDa | 96Pack |
Each for $1,690.00
|
|
|||||
UFC910024
![]() |
MilliporeSigma™
UFC910024 |
100 kDa | 24Pack |
Each for $444.79
|
![]() |
|||||
UFC910096
|
MilliporeSigma™
UFC910096 |
100 kDa | 96Pack |
Each for $1,690.00
|
|
|||||
UFC900324
|
MilliporeSigma™
UFC900324 |
3 kDa | 24Pack |
Each for $435.00
|
|
|||||
UFC910008
|
MilliporeSigma™
UFC910008 |
100 kDa | 8Pack |
Each for $166.00
|
|
|||||
UFC905024
|
MilliporeSigma™
UFC905024 |
50 kDa | 24Pack |
Each for $435.00
|
|
|||||
Description
Achieve both high sample recovery and fast sample processing. EMD Amicon™ Ultra Centrifugal Filter Units feature regenerated cellulose membranes that are ideal for protein & nucleic acid purification, concentration and desalting.
- High-recovery Ultracel™-PL regenerated cellulose membranes with MWCOs of 3, 10, 30, 50 or 100kDa
- Vertical membrane reduces polarization for ultrafast spin times (as little as 10 to 15 min.)
- Heat-sealed membrane eliminates possibility of downstream extractables
- Universal rotor compatibility
- High retentate recovery of >90%
- High concentration factors of 80- to 100-fold (minimum final concentrate volume: 200ÎĽL)
- Easy sample monitoring due to translucent housing and volume gradations
- Direct pipetter sample access eliminates processing step to recover concentrate
- Concentration of biological samples containing antigens, antibodies, enzymes, nucleic acids, or microorganisms
- Purification of macromolecular components found in tissue culture extracts or cell lysates
- Concentration of dilute or prepurified proteins from column eluents
- Desalting and buffer exchange
Specifications
Single-use | |
Non-sterile | |
Protein purification, protein concentration and desalting | |
Two microcentrifuge tubes | |
15 mL | |
29.7 mm |
High-recovery Ultracel-PL Regenerated Cellulose | |
7.6 cm2 | |
Regenerated Cellulose (RC) | |
Centrifugal Filters | |
121 mm |