Learn More
8-Bromoadenosine-3',5'-cyclic monophosphate
CAS: 23583-48-4 | C10H11BrN5O6P | 408.11 g/mol
Supplier: Thermo Scientific Chemicals J64469MF
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Environmental benefits include:
- Renewable Energy
Specifications
8-Bromoadenosine-3',5'-cyclic monophosphate | |
254°C | |
C10H11BrN5O6P | |
50 mg | |
bcamp, 8-bromo-cyclic amp, 8-br-camp, 8-bromoadenosine 3',5'-cyclic monophosphate, 8-bromo-cyclic 3',5'-amp, 8-bromo-camp, 8-bromo cyclic adenosine monophosphate, 8-bromoadenosine 3',5'-monophosphate, 8-bromoadenosine cyclic 3',5'-phosphate, cyclic 8-bromoadenosine 3',5'-monophosphate | |
NC1=C2N=C(Br)N([C@@H]3O[C@@H]4COP(O)(=O)O[C@H]4[C@H]3O)C2=NC=N1 | |
408.11 | |
CHEBI:64211 | |
Powder |
23583-48-4 | |
Brown | |
MFCD00075580 | |
Hygroscopic | |
DVKQVRZMKBDMDH-UUOKFMHZSA-N | |
(4aR,6R,7R,7aS)-6-(6-amino-8-bromopurin-9-yl)-2-hydroxy-2-oxo-4a,6,7,7a-tetrahydro-4H-furo[3,2-d][1,3,2]dioxaphosphinin-7-ol | |
32014 | |
408.11 |
Chemical Identifiers
23583-48-4 | |
408.11 | |
DVKQVRZMKBDMDH-UUOKFMHZSA-N | |
32014 | |
(4aR,6R,7R,7aS)-6-(6-amino-8-bromopurin-9-yl)-2-hydroxy-2-oxo-4a,6,7,7a-tetrahydro-4H-furo[3,2-d][1,3,2]dioxaphosphinin-7-olShow More |
C10H11BrN5O6P | |
MFCD00075580 | |
bcamp, 8-bromo-cyclic amp, 8-br-camp, 8-bromoadenosine 3',5'-cyclic monophosphate, 8-bromo-cyclic 3',5'-amp, 8-bromo-camp, 8-bromo cyclic adenosine monophosphate, 8-bromoadenosine 3',5'-monophosphate, 8-bromoadenosine cyclic 3',5'-phosphate, cyclic 8-bromoadenosine 3',5'-monophosphateShow More | |
CHEBI:64211 | |
NC1=C2N=C(Br)N([C@@H]3O[C@@H]4COP(O)(=O)O[C@H]4[C@H]3O)C2=NC=N1Show More |
Safety and Handling
EINECSNumber : 245-760-6
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only