missing translation for 'onlineSavingsMsg'
Learn More

8-Bromoadenosine-3',5'-cyclic monophosphate

Catalog No. p-7042938
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ64469MF 50 mg
AAJ64469MC 100 mg
AAJ64469MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ64469MF Supplier Thermo Scientific Chemicals Supplier No. J64469MF

CAS: 23583-48-4 | C10H11BrN5O6P | 408.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 23583-48-4
Molecular Formula C10H11BrN5O6P
Molecular Weight (g/mol) 408.11
MDL Number MFCD00075580
InChI Key DVKQVRZMKBDMDH-UUOKFMHZSA-N
Synonym bcamp, 8-bromo-cyclic amp, 8-br-camp, 8-bromoadenosine 3',5'-cyclic monophosphate, 8-bromo-cyclic 3',5'-amp, 8-bromo-camp, 8-bromo cyclic adenosine monophosphate, 8-bromoadenosine 3',5'-monophosphate, 8-bromoadenosine cyclic 3',5'-phosphate, cyclic 8-bromoadenosine 3',5'-monophosphate
PubChem CID 32014
ChEBI CHEBI:64211
IUPAC Name (4aR,6R,7R,7aS)-6-(6-amino-8-bromopurin-9-yl)-2-hydroxy-2-oxo-4a,6,7,7a-tetrahydro-4H-furo[3,2-d][1,3,2]dioxaphosphinin-7-ol
SMILES NC1=C2N=C(Br)N([C@@H]3O[C@@H]4COP(O)(=O)O[C@H]4[C@H]3O)C2=NC=N1

Specifications

Melting Point 254°C
Color Brown
Quantity 50 mg
Sensitivity Hygroscopic
Formula Weight 408.11
Physical Form Powder
Chemical Name or Material 8-Bromoadenosine-3',5'-cyclic monophosphate
EINECSNumber 245-760-6
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only