Learn More
4-Iodophenol, 99%
CAS: 540-38-5 | C6H5IO | 220.01 g/mol
Supplier: Thermo Scientific Chemicals 122390250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Environmental benefits include:
- Renewable Energy
Specifications
4-Iodophenol | |
89.0°C to 94.0°C | |
287.0°C | |
98.5% min. (GC) | |
C6H5IO | |
MFCD00002327 | |
06,208 | |
p-iodophenol, phenol, 4-iodo, phenol, p-iodo, 4-hydroxyiodobenzene, p-hydroxyiodobenzene, 4-hydroxyphenyl iodide, 4-iodo-phenol, unii-bh194bak0b, ccris 668, bh194bak0b | |
VSMDINRNYYEDRN-UHFFFAOYSA-N | |
4-iodophenol | |
10894 | |
220.01 | |
Crystalline Powder or Crystals |
540-38-5 | |
Beige to Brown or Pink | |
Authentic | |
Glass bottle | |
IC6H4OH | |
25 g | |
15,5079 | |
Solubility in water: slightly soluble. Other solubilities: freely soluble in alcohol and ether | |
OC1=CC=C(I)C=C1 | |
220.01 | |
CHEBI:43521 | |
99% |
Chemical Identifiers
540-38-5 | |
220.01 | |
VSMDINRNYYEDRN-UHFFFAOYSA-N | |
10894 | |
4-iodophenol |
C6H5IO | |
MFCD00002327 | |
p-iodophenol, phenol, 4-iodo, phenol, p-iodo, 4-hydroxyiodobenzene, p-hydroxyiodobenzene, 4-hydroxyphenyl iodide, 4-iodo-phenol, unii-bh194bak0b, ccris 668, bh194bak0bShow More | |
CHEBI:43521 | |
OC1=CC=C(I)C=C1 |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
GHS Signal Word: Danger
EINECSNumber : 208-745-5
RUO – Research Use Only