missing translation for 'onlineSavingsMsg'
Learn More

4-Iodophenol, 99%

Catalog No. p-4509698
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC122390100 10 g Glass bottle
AC122390250 25 g Glass bottle
AC122391000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC122390100 Supplier Thermo Scientific Chemicals Supplier No. 122390100
Only null left

CAS: 540-38-5 | C6H5IO | 220.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 540-38-5
Molecular Formula C6H5IO
Molecular Weight (g/mol) 220.01
MDL Number MFCD00002327
InChI Key VSMDINRNYYEDRN-UHFFFAOYSA-N
Synonym p-iodophenol, phenol, 4-iodo, phenol, p-iodo, 4-hydroxyiodobenzene, p-hydroxyiodobenzene, 4-hydroxyphenyl iodide, 4-iodo-phenol, unii-bh194bak0b, ccris 668, bh194bak0b
PubChem CID 10894
ChEBI CHEBI:43521
IUPAC Name 4-iodophenol
SMILES OC1=CC=C(I)C=C1

Specifications

Melting Point 89.0°C to 94.0°C
Color Beige to Brown or Pink
Boiling Point 287.0°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula IC6H4OH
Quantity 10 g
Beilstein 06,208
Merck Index 15,5079
Solubility Information Solubility in water: slightly soluble. Other solubilities: freely soluble in alcohol and ether
Formula Weight 220.01
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 4-Iodophenol
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
EINECSNumber 208-745-5

RUO – Research Use Only