missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4'-Methylenebis(2-chloroaniline) 90.0+%, TCI America™
$59.65 - $262.83
Chemical Identifiers
| CAS | 101-14-4 |
|---|---|
| Molecular Formula | C13H12Cl2N2 |
| Molecular Weight (g/mol) | 267.15 |
| MDL Number | MFCD00047829 |
| InChI Key | IBOFVQJTBBUKMU-UHFFFAOYSA-N |
| Synonym | 4,4'-methylenebis 2-chloroaniline, moca, quodorole, cyanaset, bisamine, dacpm, mboca, curalin m, diamet kh, methylenebis chloroaniline |
| PubChem CID | 7543 |
| ChEBI | CHEBI:28124 |
| IUPAC Name | 4-[(4-amino-3-chlorophenyl)methyl]-2-chloroaniline |
| SMILES | NC1=CC=C(CC2=CC=C(N)C(Cl)=C2)C=C1Cl |
Chemical Identifiers
| 101-14-4 | |
| 267.15 | |
| IBOFVQJTBBUKMU-UHFFFAOYSA-N | |
| 7543 | |
| 4-[(4-amino-3-chlorophenyl)methyl]-2-chloroaniline |
| C13H12Cl2N2 | |
| MFCD00047829 | |
| 4,4'-methylenebis 2-chloroaniline, moca, quodorole, cyanaset, bisamine, dacpm, mboca, curalin m, diamet kh, methylenebis chloroaniline | |
| CHEBI:28124 | |
| NC1=CC=C(CC2=CC=C(N)C(Cl)=C2)C=C1Cl |
Specifications
| 101-14-4 | |
| Yellow | |
| MFCD00047829 | |
| 3077 | |
| IBOFVQJTBBUKMU-UHFFFAOYSA-N | |
| 4-[(4-amino-3-chlorophenyl)methyl]-2-chloroaniline | |
| 7543 | |
| 267.15 | |
| Crystalline Powder |
| 108°C | |
| C13H12Cl2N2 | |
| 25 g | |
| 4,4'-methylenebis 2-chloroaniline, moca, quodorole, cyanaset, bisamine, dacpm, mboca, curalin m, diamet kh, methylenebis chloroaniline | |
| NC1=CC=C(CC2=CC=C(N)C(Cl)=C2)C=C1Cl | |
| 267.15 | |
| CHEBI:28124 | |
| ≥90.0% (HPLC) | |
| 4,4′-Methylenebis(2-chloroaniline) |
Safety and Handling
EINECSNumber : (4)-0095&(4)-0096&(4)-0275
RTECSNumber : CY1050000
TSCA : Yes