missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Methylenebis(2-chloroaniline) 90.0+%, TCI America™

Supplier:  TCI America M0609500G

 View more versions of this product

Catalog No. M0609500G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

4,4′-Methylenebis(2-chloroaniline)
108°C
C13H12Cl2N2
500 g
4,4'-methylenebis 2-chloroaniline, moca, quodorole, cyanaset, bisamine, dacpm, mboca, curalin m, diamet kh, methylenebis chloroaniline
NC1=CC=C(CC2=CC=C(N)C(Cl)=C2)C=C1Cl
267.15
CHEBI:28124
≥90.0% (HPLC)
101-14-4
Yellow
MFCD00047829
3077
IBOFVQJTBBUKMU-UHFFFAOYSA-N
4-[(4-amino-3-chlorophenyl)methyl]-2-chloroaniline
7543
267.15
Crystalline Powder

Chemical Identifiers

101-14-4
267.15
IBOFVQJTBBUKMU-UHFFFAOYSA-N
7543
4-[(4-amino-3-chlorophenyl)methyl]-2-chloroaniline
C13H12Cl2N2
MFCD00047829
4,4'-methylenebis 2-chloroaniline, moca, quodorole, cyanaset, bisamine, dacpm, mboca, curalin m, diamet kh, methylenebis chloroaniline
CHEBI:28124
NC1=CC=C(CC2=CC=C(N)C(Cl)=C2)C=C1Cl
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (4)-0095&(4)-0096&(4)-0275

RTECSNumber : CY1050000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions