missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Phenoxypropionic Acid 98.0+%, TCI America™
Supplier: TCI America P0318500G
Specifications
| 2-Phenoxypropionic Acid | |
| 116°C | |
| C9H10O3 | |
| 500 g | |
| SXERGJJQSKIUIC-UHFFFAOYSA-N | |
| 2-phenoxypropanoic acid | |
| 13658 | |
| ≥98.0% (GC,T) |
| 940-31-8 | |
| White | |
| MFCD00002643 | |
| 2-phenoxypropionic acid, alpha-methylphenoxyacetic acid, propanoic acid, 2-phenoxy, alpha-phenoxypropionic acid, phenoxypropionic acid, acide phenoxy-2-propionique, propionic acid, 2-phenoxy, dl-2-phenoxypropionic acid, 2rs-2-phenoxypropionic acid, acide phenoxy-2-propionique french | |
| CC(C(=O)O)OC1=CC=CC=C1 | |
| 166.176 | |
| 166.18 | |
| Crystalline Powder |
Chemical Identifiers
| 940-31-8 | |
| 166.176 | |
| SXERGJJQSKIUIC-UHFFFAOYSA-N | |
| 13658 | |
| CC(C(=O)O)OC1=CC=CC=C1 |
| C9H10O3 | |
| MFCD00002643 | |
| 2-phenoxypropionic acid, alpha-methylphenoxyacetic acid, propanoic acid, 2-phenoxy, alpha-phenoxypropionic acid, phenoxypropionic acid, acide phenoxy-2-propionique, propionic acid, 2-phenoxy, dl-2-phenoxypropionic acid, 2rs-2-phenoxypropionic acid, acide phenoxy-2-propionique french | |
| 2-phenoxypropanoic acid |
Safety and Handling
EINECSNumber : (3)-0746&(3)-1770&(9)-1257
RTECSNumber : UF6515000
TSCA : Yes