missing translation for 'onlineSavingsMsg'
Learn More

2-Phenoxypropionic Acid 98.0+%, TCI America™

Catalog No. p-7134554
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P031825G 25 g
P0318500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P031825G Supplier TCI America Supplier No. P031825G
Only null left

Chemical Identifiers

CAS 940-31-8
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00002643
InChI Key SXERGJJQSKIUIC-UHFFFAOYSA-N
Synonym 2-phenoxypropionic acid, alpha-methylphenoxyacetic acid, propanoic acid, 2-phenoxy, alpha-phenoxypropionic acid, phenoxypropionic acid, acide phenoxy-2-propionique, propionic acid, 2-phenoxy, dl-2-phenoxypropionic acid, 2rs-2-phenoxypropionic acid, acide phenoxy-2-propionique french
PubChem CID 13658
IUPAC Name 2-phenoxypropanoic acid
SMILES CC(C(=O)O)OC1=CC=CC=C1

Specifications

Melting Point 116°C
Color White
Quantity 25 g
Formula Weight 166.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Phenoxypropionic Acid
EINECSNumber (3)-0746&(3)-1770&(9)-1257
RTECSNumber UF6515000
TSCA Yes