CAS RN 519-32-4
CAS RN 519-32-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Caffeine Impurity C Pharmaceutical Secondary Standard, MilliporeSigmaâ„¢ Supelcoâ„¢
Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards