missing translation for 'onlineSavingsMsg'
Learn More

Caffeine Impurity C Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014422
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014422 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014422 Supplier Merck Emd Millipore Supplier No. PHR199950MG

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 519-32-4
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C8H10N4O2
MDL Number MFCD00022834
Synonym 1,3,9-Trimethylxanthine; 1,3,9-TRIMETHYL-3,9-DIHYDRO -1 H-PURINE-2,6-DIONE; 2,6-Dihydroxy-1,3,9-trimethylpurine; Isocaffeine
Type Certified Reference Material
Quantity 50 mg
Formula Weight 194.19
Grade Certified Reference Material