Learn More
Zinc wire, 1.0mm (0.04in) dia, 99.9997% (metals basis)
CAS: 7440-66-6 | Zn | 65.38 g/mol
Supplier: Thermo Scientific Chemicals 011361G5
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Zinc wire | |
7440-66-6 | |
Zn | |
5 m | |
HCHKCACWOHOZIP-UHFFFAOYSA-N | |
zinc | |
23994 | |
99.9997% | |
≈5.60g/m |
1.0mm (0.04 in.) dia. | |
(metals basis) | |
MFCD00011291 | |
dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust | |
[Zn] | |
65.38 | |
CHEBI:30185 | |
Odorless | |
Wire |
Chemical Identifiers
7440-66-6 | |
65.38 | |
HCHKCACWOHOZIP-UHFFFAOYSA-N | |
23994 | |
zinc |
Zn | |
MFCD00011291 | |
dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dustShow More | |
CHEBI:30185 | |
[Zn] |
Safety and Handling
EINECSNumber : 231-175-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only