missing translation for 'onlineSavingsMsg'
Learn More

Zinc wire, 1.0mm (0.04in) dia, 99.9997% (metals basis)

Catalog No. p-7048730
Change view
Click to view available options
Quantity:
1 m
5 m
20 m
Catalog No. Quantity
AA11361G1 1 m
AA11361G5 5 m
AA11361G8 20 m
3 options

Catalog No. AA11361G1

Supplier: Thermo Scientific Chemicals 011361G1

Only null left

CAS: 7440-66-6 | Zn | 65.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-66-6
Molecular Formula Zn
Molecular Weight (g/mol) 65.38
MDL Number MFCD00011291
InChI Key HCHKCACWOHOZIP-UHFFFAOYSA-N
Synonym dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
PubChem CID 23994
ChEBI CHEBI:30185
IUPAC Name zinc
SMILES [Zn]

Specifications

Quantity 1 m
Odor Odorless
Weight ≈5.60g/m
Form Wire
Percent Purity 99.9997%
Assay (metals basis)
Chemical Name or Material Zinc wire, 1.0mm (0.04 in.) dia.
EINECSNumber 231-175-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only