missing translation for 'onlineSavingsMsg'
Learn More

Yttrium(III) trifluoroacetate hydrate, 99.9% (REO)

CAS: 304851-95-4 | C6F9O6Y | 427.95 g/mol

Supplier:  Thermo Scientific Chemicals 04459718

 View more versions of this product

Catalog No. AA4459718


Only null left
Explore available promotions

Description

Description

A fluorinated yttrium compound for proteomics research. Used in the manufacture of ceramics, glass, laser materials, phosphor. non-ferrous alloy iron and steel, and the additives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Greener Choice Claims
  • Environmental benefits include:
    • Recyclable
Specifications
Show More
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only