missing translation for 'onlineSavingsMsg'
Learn More

Yttrium(III) trifluoroacetate hydrate, 99.9% (REO)

Catalog No. p-7059431
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4459709 10 g
AA4459718 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4459709 Supplier Thermo Scientific Chemicals Supplier No. 04459709
Only null left

CAS: 304851-95-4 | C6F9O6Y | 427.95 g/mol

A fluorinated yttrium compound for proteomics research. Used in the manufacture of ceramics, glass, laser materials, phosphor. non-ferrous alloy iron and steel, and the additives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 304851-95-4
Molecular Formula C6F9O6Y
Molecular Weight (g/mol) 427.95
MDL Number MFCD05663780 MFCD00192577
InChI Key NPATVYPXYMSPKD-UHFFFAOYSA-K
Synonym yttrium trifluoroacetate
PubChem CID 22836421
IUPAC Name 2,2,2-trifluoroacetic acid;yttrium;hydrate
SMILES [Y+3].[O-]C(=O)C(F)(F)F.[O-]C(=O)C(F)(F)F.[O-]C(=O)C(F)(F)F

Specifications

Physical Form Crystalline
Quantity 10 g
Solubility Information Insoluble in water.
Formula Weight 427.95 (Anhydrous)
Percent Purity 99.9%
Assay (REO)
Chemical Name or Material Yttrium (III) trifluoroacetate hydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only