Learn More
DWK Life Sciences Wheaton™ LDPE Dropping Bottles
Excellent chemical resistance
$226.97 - $4170.18
Catalog Number | Mfr. No. | Capacity (Metric) | Closure Size | Color | Quantity | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Closure Size | Color | Quantity | Price | Quantity | |||
03-439-71H
|
DWK Life Sciences
W242831 |
3 mL | 8-425 mm | White | 100/Cs. |
Case of 100 Each for $226.97
|
|
|||
03-439-72G
|
DWK Life Sciences
W242822 |
6 mL | 13-425 mm | Natural | 100/Cs. |
Case of 100 Each for $326.94
|
|
|||
03-439-72F
|
DWK Life Sciences
W242823 |
7 mL | 15-415 mm | Natural | 100/Cs. |
Case of 100 Each for $243.39
|
|
|||
03-439-75
|
DWK Life Sciences
W242825A |
15 mL | 15-415 mm | Natural | 100/Cs. |
Case of 1000 Each for $2,987.00
|
|
|||
03-439-74G
|
DWK Life Sciences
W242826A |
30 mL | 20-410 mm | Natural | 1000/Cs. |
Case of 1000 Each for $4,170.18
|
|
|||
03-439-72B
|
DWK Life Sciences
W242827 |
60 mL | 20-410 mm | Natural | 100/Cs. |
Case of 100 Each for $529.58
|
|
|||
03-439-71B
|
DWK Life Sciences
W242837 |
60 mL | 20-410 mm | White | 100/Cs. |
Case of 100 Each for $492.73
|
|
|||
Description
- Manufactured to rigid ISO and quality standards
- Excellent alternative to pipetting
- Double bagged and twist-tied to reduce particulate
- Methods of sterilization: gas and gamma
- Lot controlled
Order Info
Dropper tips and caps must be ordered separately.
Specifications
LDPE | |
No | |
Not Autoclavable |