missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Quantitative Hardened Ashless Filter Paper - Acid hardened to reduce ash to extremely low level.

Acid hardened to reduce ash to extremely low level.

Supplier:  Cytiva 1541400

Numéro de catalogue. 09940001


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

  • High wet strength
  • Chemically resistant with tough surface
  • Enables fast filtration of coarse particles and gelatinous precipitates
Spécifications

Spécifications

3 s/100 mL/in2
Hardened Ashless
Cotton Linters
Typical applications include fiber in animal foodstuffs, gelatine in milk and cream, chloride in cement, and chloride and phosphorous in coal and coke.
100/Pk.
Quantitative Filter Paper
Grade 541 Quantitative Filter Paper Hardened Ashless, circle, 400 mm
Disc
22 μm
Grade 541
155 μm
400 mm
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions