Learn More
Cytiva Whatman™ Puradisc H-PTFE Syringe Filters
Cytiva Life Sciences Whatman™ Puradisc H-PTFE Syringe Filters can be used for both aqueous and aggressive organic solvents
$176.55 - $4559.16
Specifications
Filter Type Hydrophilic
Material (Membrane) PTFE (Hydrophilic)
Sterility Non-sterile
For Use With (Application) Aqueous Solvents, Aggressive Organic Solvents, Rapid Syringe Filtration, HPLC, uHPLC
Housing Material Pigment Free Polypropylene
View More Specs
Products 12
Catalog Number Mfr. No. Pore Size Diameter (Metric) Quantity Price Quantity  
Catalog Number Mfr. No. Pore Size Diameter (Metric) Quantity Price Quantity  
09924601
View Documents Promotion Details
Cytiva
67721302
0.2 ÎĽm 13 mm 100/Pk.
Pack of 100 Each for $350.46
Only null left
09924602
View Documents
Cytiva
67731302
0.2 ÎĽm 500/Pk.
Pack of 500 Each for $1,317.19
Only null left
09924603
View Documents
Cytiva
67741302
0.2 ÎĽm 13 mm 2000/Pk.
Pack of 2000 Each for $4,559.16
Only null left
09924604
View Documents
Cytiva
67721304
0.45 ÎĽm 13 mm 100/Pk.
Pack of 100 Each for $350.46
Only null left
09924605
View Documents
Cytiva
67731304
0.45 ÎĽm 13 mm 500/Pk.
Pack of 500 Each for $1,317.19
Only null left
09924606
View Documents
Cytiva
67741304
0.45 ÎĽm 13 mm 2000/Pk.
Pack of 2000 Each for $4,304.21
Only null left
09924595
View Documents Promotion Details
Cytiva
67722502
0.2 ÎĽm 25 mm 50/Pk.
Pack of 50 Each for $176.55
Only null left
09924596
View Documents
Cytiva
67732502
0.2 ÎĽm 25 mm 200/Pk.
Pack of 200 Each for $521.87
Only null left
09924597
View Documents
Cytiva
67742502
0.2 ÎĽm 25 mm 1000/Pk.
Pack of 1000 Each for $2,298.02
Only null left
09924598
View Documents
Cytiva
67722504
0.45 ÎĽm 50/Pk.
Pack of 50 Each for $176.55
Only null left
09924599
View Documents
Cytiva
67732504
0.45 ÎĽm 25 mm 200/Pk.
Pack of 200 Each for $521.87
Only null left
09924600
View Documents
Cytiva
67742504
0.45 ÎĽm 25 mm 1000/Pk.
Pack of 1000 Each for $2,168.91
Only null left
Description
Description
  • Whatman™ Puradisc H-PTFE syringe filter contains a hydrophilic membrane that is chemically stable and inert
  • The H-PTFE membrane exhibits low protein binding and can be used for both aqueous and aggressive organic solvents.
Specifications
Specifications
Hydrophilic
Non-sterile
Pigment Free Polypropylene
Female Luer Lock
5.2 bar (75 psi)
PTFE (Hydrophilic)
Aqueous Solvents, Aggressive Organic Solvents, Rapid Syringe Filtration, HPLC, uHPLC
Hydrophilic
Male Luer
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'