missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ Anotop™ 10 Sterile Syringe Filters
Hold-up volume: <30ÎĽL (10mm size), <200ÎĽL (25mm)
Supplier: Cytiva 68091112
Description
Hold-up volume: <30ÎĽL (10mm size), <200ÎĽL (25mm)
Similar to Anotop filters, but include binder-free, glass microfiber prefilter with Anotop™ inorganic membrane in a polypropylene housing.
Glass Microfiber Prefilter
- Increases throughput
- Maintains membrane performance
- Reduces need for centrifuging before filtering, sequential filtering, or refiltering
- Permits use of 0.1ÎĽm pore size Anotop Plus filters to remove mycoplasma contaminants from tissue culture
- Hold-up volume: <30ÎĽL (10mm size), <200ÎĽL (25mm)
- Pressure limit, 7kg/cm (100psi)
Greener Choice Claims
- Environmental benefits include:
- Renewable Energy
Specifications
Single-use | |
Alumina-based Anopore | |
Sterile | |
HPLC aqueous/organic sample preparation; cold sterilization of growth media; solvent filtration for spectroanalysis; phage and virus filtration; liposome extrusion; removal of high molecular weight proteins or polymers; colloid removal from deionized water | |
Polypropylene (PP) | |
Male Luer | |
50/Pk. | |
45°C | |
Syringe Filter | |
10 mm |
Anapore | |
0.1 ÎĽm | |
0.78 cm2 | |
<20 ÎĽL | |
Female Luer Lock | |
6.9 bar (100 psi) | |
113°F | |
Polypropylene | |
<30 ÎĽL (Hold-Up Volume 10 mm Size), <200 ÎĽL (Hold-Up Volume 25 mm Size) |