missing translation for 'onlineSavingsMsg'
Learn More

TCR V alpha 3.2 Monoclonal Antibody (RR3-16), FITC, eBioscienceā„¢

TCR V alpha 3.2 Monoclonal Antibody (RR3-16), FITC, eBioscienceā„¢

Supplier:  Thermo Fisher Scientific 11579980

 View more versions of this product

Catalog No. 501122138


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

TCR V alpha 3.2
Monoclonal
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.2
Rat
25 μg
Primary
4°C, store in dark, DO NOT FREEZE!
Liquid
Flow Cytometry
RR3-16
FITC
TCR V alpha3.2; TCRV alpha 3.2; TCRV alpha3.2; Va3.2; Valpha3.2
Affinity chromatography
RUO
Mouse
Antibody
IgG2b Īŗ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions