Learn More
Honeywell™ Uvex™ Seismic™ Sealed Eyewear with Reading Magnifiers
Delivering unmatched protection against impact, wind, dust and debris, Uvex Seismic offers a range of diopter strengths for workers needing increased magnification for reading, inspection and other fine detail work.
Supplier: Honeywell™ S0661X
Description
The wraparound design provides exceptional coverage while precise placement of the magnifiers allow for distortion-free vision. Uvex Seismic Reading Magnifiers: powerful eyewear for today's demanding work environments.
- Superior coverage ideal for dirty, high-particulate work environments such as grinding, chipping, shaving and sanding.
- Available in Clear with Uvextra™ Anti-fog coating and three diopter strengths: +1.5. +2.0, and +2.5 magnifications.
- Superior anti-fog coating, coupled with top and bottom indirect ventilation, minimizes fogging to keep workers safe and comfortable all day.
- Flexible design can be worn with temples or with headband as conditions change.
- Soft, flexible nasal pads conform to a variety of facial profiles and minimize slippage.
- Soft, high-quality face cushioning and padded temple tips offer all-day comfort.
- Highly adjustable flame-resistant headband provides excellent fit.
- Replaceable flame-resistant, cushion-lined subframe and headband extend the life of the eyewear.
- High impact protection certified to ANSI Z87+(High Impact) and tested by Honeywell to meet the requirements of the CSA Z94.3 standard. (ANSI-certified for use with and without cushion-lined frame).
Greener Choice Claims
- Environmental benefits include:
- Renewable Energy
Specifications
Anti-fog | |
Sealed Eyewear with Reading Magnifiers | |
+1.50 | |
Polycarbonate |
Clear | |
Black | |
Polycarbonate | |
UV400 |