Learn More
Thermo Scientificâ„¢ Unifilter Direct-Connection Filter System Components
Minimize downtime with these easily replaceable filter system components.
$230.23 - $266.45
Specifications
For Use With (Equipment) Unifilter Direct-Connection HPLC Filter Systems
View More Specs
Products 2
Catalog Number Mfr. No. Diameter (Metric) Outer Description Type Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Outer Description Type Price Quantity  
27000
View Documents
Thermo Scientificâ„¢
CMD27000
4.0/4.6 mm Unifilter Direct-Connection Filter Holder; 4.0/4.6 mm Unifilter Direct Connection Holder
Each for $266.45
Only null left
27002
View Documents
Thermo Scientificâ„¢
27002
3.0/2.1 mm Unifilter Direct-Connection Filter Holder; 2.1/3.0 mm Unifilter Direct Connection Holder
Each for $230.23
Only null left
Description
Description
Minimize downtime. Thermo Scientificâ„¢ Unifilter Direct-Connection Filter System Components are easily replaced.
  • Replaceable 0.5μm drop-in filter enhances column lifetime and improved performance
  • Holder attached directly to the inlet of analytical system for maximum convenience
Specifications
Specifications
Unifilter Direct-Connection HPLC Filter Systems
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.