missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triethylene Glycol Divinyl Ether (stabilized with KOH) 98.0+%, TCI America™

$136.06 - $411.97
Chemical Identifiers
CAS | 765-12-8 |
---|---|
Molecular Formula | C10H18O4 |
Molecular Weight (g/mol) | 202.25 |
MDL Number | MFCD00074891 |
InChI Key | CYIGRWUIQAVBFG-UHFFFAOYSA-N |
PubChem CID | 3033992 |
IUPAC Name | 1,2-bis(2-ethenoxyethoxy)ethane |
SMILES | C=COCCOCCOCCOC=C |
Chemical Identifiers
765-12-8 | |
202.25 | |
CYIGRWUIQAVBFG-UHFFFAOYSA-N | |
1,2-bis(2-ethenoxyethoxy)ethane |
C10H18O4 | |
MFCD00074891 | |
3033992 | |
C=COCCOCCOCCOC=C |
Specifications
765-12-8 | |
126°C | |
MFCD00074891 | |
CYIGRWUIQAVBFG-UHFFFAOYSA-N | |
1,2-bis(2-ethenoxyethoxy)ethane | |
3033992 | |
≥98.0% (GC) | |
Triethylene Glycol Divinyl Ether (stabilized with KOH) |
Colorless | |
C10H18O4 | |
25 g | |
C=COCCOCCOCCOC=C | |
202.25 | |
202.25 | |
Liquid |
Safety and Handling
EINECSNumber : (7)-1321
RTECSNumber : XF0877100
TSCA : Yes