missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triethylene Glycol Divinyl Ether (stabilized with KOH) 98.0+%, TCI America™
Supplier: TCI America T3460100G
Specifications
| Triethylene Glycol Divinyl Ether (stabilized with KOH) | |
| Colorless | |
| C10H18O4 | |
| 100 g | |
| C=COCCOCCOCCOC=C | |
| 202.25 | |
| 202.25 | |
| Liquid |
| 765-12-8 | |
| 126°C | |
| MFCD00074891 | |
| CYIGRWUIQAVBFG-UHFFFAOYSA-N | |
| 1,2-bis(2-ethenoxyethoxy)ethane | |
| 3033992 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 765-12-8 | |
| 202.25 | |
| CYIGRWUIQAVBFG-UHFFFAOYSA-N | |
| 1,2-bis(2-ethenoxyethoxy)ethane |
| C10H18O4 | |
| MFCD00074891 | |
| 3033992 | |
| C=COCCOCCOCCOC=C |
Safety and Handling
EINECSNumber : (7)-1321
RTECSNumber : XF0877100
TSCA : Yes