Learn More
Titanium boride, 99.5% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04455818
Description
Used as a cathode material in aluminum smelting and can be shaped by electrical discharge machining. Specialized applications in such areas as impact resistant armor, cutting tools, crucibles, neutron absorbers and wear resistant coatings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Titanium boride | |
12045-63-5 | |
50 g | |
B2Ti | |
titanium boride, | |
QYEXBYZXHDUPRC-UHFFFAOYSA-N | |
boron;titanium | |
131664302 | |
4.38 g/mL |
3225°C | |
Powder | |
99.5% (metals basis) | |
MFCD00049595 | |
Insoluble in water. | |
[B].[B].[Ti] | |
69.487 | |
69.5 |
Chemical Identifiers
12045-63-5 | |
69.487 | |
QYEXBYZXHDUPRC-UHFFFAOYSA-N | |
131664302 | |
[B].[B].[Ti] |
B2Ti | |
MFCD00049595 | |
titanium boride, | |
boron;titanium |
Safety and Handling
EINECSNumber : 234-961-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only