missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Titanium boride, 99.5% (metals basis), Thermo Scientific Chemicals
Description
Used as a cathode material in aluminum smelting and can be shaped by electrical discharge machining. Specialized applications in such areas as impact resistant armor, cutting tools, crucibles, neutron absorbers and wear resistant coatings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 3225°C |
| Physical Form | Powder |
| Quantity | 10 g |
| Assay Percent Range | 99.5% (metals basis) |
| Solubility Information | Insoluble in water. |
| Formula Weight | 69.5 |
| Density | 4.38 |
| Chemical Name or Material | Titanium boride |
Safety and Handling
| EINECSNumber | 234-961-4 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only