missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TG Rabbit anti-Human, Unconjugated, Clone: TGB/1968R, Abnova™
Rabbit Monoclonal Antibody.
Supplier: Abnova Corporation RAB00640
Description
TG Monoclonal antibody specifically detects TG in Human samples. It is validated for IHC-P.Greener Choice Claims
- Environmental benefits include:
- Recyclable
Specifications
TG | |
Monoclonal | |
Unconjugated | |
TG | |
Rabbit | |
100 ÎĽg | |
7038 | |
Store at -20 to -80°C | |
IgG |
Immunohistochemistry (Paraffin) | |
TGB/1968R | |
thyroglobulin | |
AITD3, TGN | |
Recombinant protein corresponding to full-length human TG | |
Primary | |
Human | |
Liquid |