missing translation for 'onlineSavingsMsg'
Learn More

Texwipe™ VersaWipe™ Wipers

For ISO Class 6 to 7 (Class 1000 to 10,000) applications

Supplier:  Texwipe™ TX622

 View more versions of this product

Catalog No. 19-130-3863


Only null left
Explore available promotions

Description

Description

Highly absorbent, durable wipers made of a nonwoven blend of 55% cellulose and 45% polyester.

  • Blend of 55% cellulose and 45% polyester offers low extractable levels and particle count
  • Economical wiper is ideal general wiping and spill control in controlled environments
  • Compatible with solvents
  • Contain no chemical binders
  • Excellent wet and dry strength
  • Autoclavable

Order Info

12 x 12 in. wipers available 10 bags/case (150 wipers/bag).

Specifications

Specifications

Dry Cleanroom Wiper
30.5 cm
30.5 cm
Autoclavable
12 x 12 in.
12 in.
12 in.
Polyester Cellulose
60
For ISO Class 6 to 7 (Class 1000 to 10000) cleanroom and general wiping and spill control applications
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions